Squishy Circuits, Standard Kit
# de catalogue 470317-842
Fournisseur: SQUISHY CIRCUITS STORE LLC SE
Spécifications
- Description:Squishy Circuits Standard Kit
- Cat. no.:470317-842
Spécifications
A propos de cet article
The standard kit includes everything you need to get started with the more advanced Squishy Circuits projects. Easy and fun!
Breakthrough your limitations by using your creativity and engineering skills to make your projects ground-breaking and one of a kind with LEDs, buzzers, and insulating dough.
Delivery: Kit includes 1×Battery holder, 1×Piezoelectric buzzer, 25×LEDs (Five each red, green, white, yellow and blue), 1 to 3.5 Oz. Container of Insulating Dough, and 3 to 3.5 Oz. Containers of Conductive Dough (in Red, Green, and Blue).
Customers Who Bought This Also Bought