Squishy Circuits, Lite Kit
# de catalogue 470317-840
Fournisseur: SQUISHY CIRCUITS STORE LLC SE
Spécifications
- Description:Squishy Circuits Lite Kit
- Cat. no.:470317-840
Spécifications
A propos de cet article
Uses conductive play dough and insulating play dough to teach electrical circuits basics in a fun, hands-on way.
The lite kit is the most basic kit and includes everything that you need to make your first Squishy Circuits light up quickly and easily.
It is ideal for classrooms, parties, or events where every participant needs their own battery pack and wants to get started building right away.
Delivery: Kit includes 1×Battery holder, 1×Piezoelectric buzzer, 25×LEDs (5 each red, green, white, yellow and blue), 1 to 3.5 Oz. Container of Insulating Dough, and 3 to 3.5 Oz. Containers of Conductive Dough (in Red, Green, and Blue).
Customers Who Bought This Also Bought