SP Wilmad-LabGlass 4-Neck Molded Reaction Vessel Covers, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
These borosilicate glass 4-neck reaction vessel lids/covers have a contact surface that is finely ground to provide a tight seal.
- LG-8081 is designed for use with LG-8080 cylindrical reaction vessels
- LG-8083 is designed for use with LG-8082 vessels
Featuring a flat flange, these pressed lids have a center neck and three side necks with outer standard taper joints. Use LG-7316 clamps.
Customers Who Bought This Also Bought