Canadian Rock and Mineral Collection
# de catalogue 470315-520
Fournisseur: Avantor
Spécifications
- Description:Canadian Collection of Rocks and Minerals
- Cat. no.:470315-520
Spécifications
A propos de cet article
Comprehensive set of rocks and minerals highlighting the economic resources of Canada.
This thorough set contains 40 large samples of the rocks and minerals of Canada, numbered in a compartmentalized box with identification key. Perfect for the pan-Canadian curriculum guide and new text titles in Canada.
Delivery: Includes seven sedimentary rocks, 7 igneous rocks, 6 metamorphic rocks, 7 metallic minerals, 13 non-metallic minerals. Size of each 1×1 to 1×2"
Customers Who Bought This Also Bought