Corepoint Scientific™ Refrigerators, Horizon Scientific
Supplier: Horizon Scientific
Certificates
About this item
These compact laboratory refrigerators offer professional cold storage for scientific and clinical settings.
- Microprocessor temperature control
- Integrated temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
Units come equipped with forced draft circulation and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76335-186 and 76335-188
Ordering information: All units come with a three year parts and labor warranty, plus an additional two year compressor parts warranty.
Specifications
- Catalog No:
- 76335-188
- 76335-186
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Cat. No.:
- 76335-188
- 76335-186
- Defrost:
- Manual
- Manual
- Shelves/drawers:
- 6 adjustable shelves
- 6 adjustable shelves
- Door type:
- 1 solid swing door
- 1 glass swing door
- Material:
- Steel
- Steel
- Electrical:
- 115 V, 2.3 A
- 115 V, 2.3 A
- Weight:
- 83 kg (183 lbs.)
- 83 kg (183 lbs.)
- Environmental certification:
- HFC refrigerant
- HFC refrigerant