Polywipe® Light Wipes, Contec®
# de catalogue 68300-554
Fournisseur: Contec
Small Business Enterprise
Spécifications
- Dimensions:30 x 30 cm (12 x 12")
- Color:White
- Description:Polywipe Light Wipes, flat stacked
- Model:Polywipe Light
- Fibres (>100 µm):200.9×10³/m²
- Extractables:0.087 g/m² using deionized water, 0.120 g/m² using 1,1,1-trichloroethane
- Basis weight:115 g/m²
- Particle generation (>=0,5 µm):65.0 x 10⁶/m²
- Absorbency - intrinsic:3.7 ml/g
- Ions - sodium:8 ppm
- Ions - chloride:3 ppm
- Cat. no.:68300-554
- Packaging:150/Bag, 12 Bags/Case
- Supplier No.:7070-99
Spécifications
A propos de cet article
Polywipe Light Wipes are unlaundered standard weight interlock knit polyester wipes with knife-cut edges that can be used in less critical applications.
- Recommended for ISO Class 6 to 8 environments
- Knitted 100% polyester fabric for low levels of particles and fibers
- Soft edge is ideal for use on surfaces which are vulnerable to scratching
- Cost effective for equipment wipe down and general cleaning
- Durable no run interlock knit construction keeps the fabric from unraveling
- Abrasion and chemical resistant
The durable, no-run interlock knit construction makes it ideal for equipment wipe down and general cleaning. It is available in different sizes and put-ups.
Customers Who Bought This Also Bought