Amino Labs DNA Extraction Kit
Fournisseur: AMINO LABS NORTH INCORPORATED CA SE
Certificats
A propos de cet article
Visualize the DNA you eat every day.
- Allows students to extract and visualize DNA from fruit
- Quick and easy procedure takes only 15 minutes
- Great beginner biotechnology activity
The incredible DNA extraction kit takes you through a journey of extracting the genomic DNA from the cells that make up a strawberry.
Additional items required but not included: 9 ml Isopropyl alcohol (99%), strawberry, plastic or glass cup).
Ordering information: Due to popular demand, we recommend you place your order at least a week in advance.
Delivery: Though kits will ship at ambient temperature, some components will require refrigeration upon arrival. Refer to instructions.
Customers Who Bought This Also Bought