Bone Clones® Fossil Hominid Tools set of 6
# de catalogue 470317-812
Fournisseur: BONE CLONES, INC.
Spécifications
- Description:Fossil Hominid Tools set of 6
- Cat. no.:470317-812
Spécifications
A propos de cet article
Set of 6 fossil hominid tools.
- Casting from originals that present diverse osteological characteristics, each Bone Clones® cast is hand-made from start to finish
- Rigorous standards are applied when selecting original specimens to ensure the best representation
- Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
ncludes an Acheulian Banded Flint Hand-Axe from southwest Morocco; a Mousterian Neanderthal Triangular Bi-facial Hand-Axe; a Mousterian Bi-facial ovate point on a Levallois Blade; a Mid-Acheulian Cordiform Bi-facial Hand-Axe; a Mousterian 40,000 year old scraper; and an Acheulian tool. Acheulean tools (approximately 2.0 MYA-200,000 YA) are associated with Homo erectus. Mousterian tools are dated at (100,000-40,000 YA).
Weight: 2 lbs.
BodyTextAccessories: Accessories not included.
Customers Who Bought This Also Bought