Hard Tissue Homogenizing Mix, Omni International
# de catalogue 10015-242
Fournisseur: OMNI INTERNATIONAL
Spécifications
- Volume:30 ml
- Description:Hard Tissue Homogenizing Mix (30 ml Tubes)
- Cat. no.:10015-242
- Working Volume:2 - 25 ml
- Supplier No.:19-6358
Spécifications
A propos de cet article
Bead ruptor bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient and high yield sample homogenates.
- Recommended for tough and fibrous plant material, hard tissues, seeds and feed
- Made of polypropylene
- Max. centrifugal force 5000×g
30 ml polypropylene screw cap tubes are pre-filled with 2.8 mm ceramic beads.
Ordering information: Unit includes 50 bead beating tubes pre-filled with 2.8 mm ceramic beads.
Customers Who Bought This Also Bought