ContactUsLinkComponent
Bone Clones® Homo neanderthalensis Skull La Chapelle-aux-Saints
Bone Clones® Homo neanderthalensis Skull La Chapelle-aux-Saints
# de catalogue 470317-834
Fournisseur:  BONE CLONES, INC.
Model Nean Skull La Chapelle-Aux-Saints
Bone Clones® Homo neanderthalensis Skull La Chapelle-aux-Saints
# de catalogue 470317-834
Fournisseur:  BONE CLONES, INC.
Spécifications
  • Description:
    Homo neanderthalensis Skull La Chapelle-aux-Saints
  • Size:
    9D×6W×7"H
  • Cat. no.:
    470317-834
  • Weight:
    2 lbs.
Spécifications
A propos de cet article
50000 YA. The Homo neanderthalensis Skull was discovered by A. and J. Bouyssomie and J. Bonneval in 1908 in La Chapelle-aux-Saints, France. It was the most complete Neanderthal skull found at the time.
  • Each Bone Clones® cast is hand-made from start to finish
  • Rigorous standards are applied when selecting original specimens to ensure the best representation
  • Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
Called the 'old man', this individual clearly lived a long time, having much pathology, such as the declined tooth sockets in the molar region and severe arthritis in the neck. Despite his very large brain capacity (over 1,600 cc), early descriptions by Boule declared him to be primitive, leading to the term 'Neanderthal' to be synonymous with brutality for decades. More recently, however, many consider Neanderthal to have been a highly successful and social species with many of its physical features, such as low cranial vault and projected face, due to adaptations to cold weather. This cranium is representative of many fossil remains of Neanderthals found throughout present-day Europe.
The Bone Clones® Hominid line is composed of discoveries from anatomically modern humans, archaic humans, early Homo, early hominins, and other hominids. The majority of the casts in this line have been recreated by a team of anatomical sculptors. Some are reconstructions made by anthropology professionals using fragmentary elements from original discoveries and extrapolating the missing parts from those.
BodyTextAccessories: Accessories not included.
Customers Who Bought This Also Bought