ContactUsLinkComponent
Bone Clones® Australopithecus afarensis Skull
Bone Clones® Australopithecus afarensis Skull
# de catalogue 470317-774
Fournisseur:  BONE CLONES, INC.
Model Australopithecus Afarensis Skull
Bone Clones® Australopithecus afarensis Skull
# de catalogue 470317-774
Fournisseur:  BONE CLONES, INC.
Spécifications
  • Description:
    Australopithecus afarensis Skull
  • Size:
    7.5D×6.0W×7.0"H
  • Cat. no.:
    470317-774
  • Weight:
    2 lbs.
Spécifications
A propos de cet article
2.9 to 3.6 MYA. Australopithecus afarensis is the best represented early hominid with approximately 100 fossils representing the species.
  • Each Bone Clones® cast is hand-made from start to finish
  • Rigorous standards are applied when selecting original specimens to ensure the best representation
  • Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
The australopithecines are only known from Africa; none has ever been found in Europe or Asia. They had ape-sized brains: their cranial capacity ranged from 375 to 530 cc. They had strong jaws with large teeth. Like modern gorillas, the adult males were much larger than the females. The babies probably took approximately the same length of time to grow up as a modern chimpanzee or gorilla. This full size skull was sculpted for Bone Clones® by scientific sculptor Steven Wagner.
The Bone Clones® Hominid line is composed of discoveries from anatomically modern humans, archaic humans, early Homo, early hominins, and other hominids. The majority of the casts in this line have been recreated by a team of anatomical sculptors. Some are reconstructions made by anthropology professionals using fragmentary elements from original discoveries and extrapolating the missing parts from those.
BodyTextAccessories: Accessories not included.
Customers Who Bought This Also Bought