Stomacher® 4500 Lab Blender Series, Seward
Fournisseur: Seward Laboratory System
Certificats
A propos de cet article
The robust Stomacher® 4500 is ideal for the BAM large volume testing procedures from 1000 to 4500 ml.
- Easy to clean
- Easy bag loading
- Programmable settings
Stomacher® lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples. The Stomacher® machines work by the action of twin-reciprocating paddles on the sample contained inside a sterile plastic bag.
Expanding the food testing focused blenders, the Stomacher® 4500 BAM is the perfect tool for larger volume BAM testing procedures for extraction from 375 g composite samples.
Ordering information: Machine comes with three year warranty and ten year service support.
Customers Who Bought This Also Bought