VWR® Microcentrifuge Tubes, Capless
76332-078
- Sterility:Non sterile
- Volume:1.5 ml (max. 1.7 ml)
- Color:Natural
- Cat. no.:76332-078
These microcentrifuge tubes are available without caps. Best for use in fraction collection and whenever storage of samples is not necessary.
- Rated 20000 ×g
- Graduated every 100 µl
- Polypropylene
Capless tubes are graduated and are available in natural only.
Customers Who Bought This Also Bought