A propos de cet article
The Solid.Line KBF-S climate chambers are ideal for long-term testing and accelerated stability testing of pharmaceutical products in accordance with ICH Q1A guidelines. The corrosion-free and condensation-free interior ensures exceptionally stable temperature and moisture levels, even when the chamber is fully loaded.
- Humidity regulation with capacitive humidity sensor and steam humidification
- LCD to display temperature and humidity along with additional information and alarms
- Internal data logger, measured values can be read out in open format via USB
- Units self-test for comprehensive status analysis
- Access port with silicone plug, 30 mm, left
- Class 3.1 independent temperature safety device (DIN 12880) with visual and audible temperature alarm
Units have a stainless steel inner chamber and the APT.line™ preheating chamber technology ensures air is circulated evenly and gently within the usable space, creating precise and homogeneous climate conditions. KBF-S constant climate chambers can often be used as a cost-effective alternative to environmental chambers. They feature an ethernet interface, are easy to program, make documenting test results simple and include an independent, container-based water supply.
Spécifications
- Catalog No:
- 76331-846
- 76331-842
- 76403-154
- 76403-152
- Model:
- KBF S 720UL
- KBF S 240UL
- KBF S 1020UL
- KBF S 115UL
- Temperature range:
- 0…70 °C
- 0…70 °C
- 0…70 °C
- 0…70 °C
- Humidity range (RH):
- 20 - 80 %
- 20 - 80 %
- 20 - 80 %
- 20 - 80 %
- Convection type:
- Forced convection
- Forced convection
- Forced convection
- Forced convection
- Int. W×D×H:
- 973×576×1250 mm
- 650×485×785 mm
- 976×836×1250 mm
- 600×351×483 mm
- Ext. W×D×H:
- 1250×890×1925 mm
- 925×800×1460 mm
- 1250×1145×1925 mm
- 800×650×1050 mm
- Nominal power consumption:
- 2000 W
- 1400 W
- 2000 W
- 1400 W