Corepoint Scientific™ Premier™ Hydrocarbon Manual Defrost Solid Door Freezer, Horizon Scientific
# de catalogue 76333-622
Fournisseur: Horizon Scientific
Spécifications
- Temperature Range:-15...-25 °C
- Description:Premier hydrocarbon manual defrost freezer
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:R290 Refrigerant
- Cat. No.:76333-622
- Electrical:115 V, 60 Hz, 1.96 A, 1/2 HP
- Shipping weight:84 kg (185 lbs.)
- Defrost:Manual
- Door type:1 solid swing door
- Shelves/drawers:6 fixed shelves
- Exterior W×D×H:600×619×1854 mm (23.625×24.375×73")
- Capacity:396 L (14 cu.ft.)
Spécifications
A propos de cet article
These efficient manual defrost freezers provide premier cooling for laboratory settings. They offer storage solutions for a variety of scientific and clinical applications.
- Microprocessor temperature control
- Integrated temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
Units come equipped with forced draft circulation and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76333-622
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Customers Who Bought This Also Bought