ContactUsLinkComponent
Cardionics® Speakers for SAM and PAT
Cardionics® Speakers for SAM and PAT
# de catalogue 470317-948
Fournisseur:  American 3B Scientific
Cardionics® Speakers for SAM and PAT
# de catalogue 470317-948
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Speaker
  • Cat. no.:
    470317-948
Spécifications
A propos de cet article
Share sounds with everyone in the room.
  • Works With SAM and PAT Trainers
  • Groups can hear the sounds while watching positioning
Expand your SAM or PAT Auscultation Trainer with this accessory Speaker. The speaker plugs into the trainer and broadcasts the sounds being heard through the stethoscope.
Customers Who Bought This Also Bought