3B Scientific® Image Guided Cervical Injection Trainer
# de catalogue 470318-132
Fournisseur: American 3B Scientific
Spécifications
- Description:Cervical Injection Trainer
- Cat. no.:470318-132
Spécifications
A propos de cet article
An economic alternative to cadaver practice.
- Life-Like radiopacity for Realistic X-Ray Images
- Realistic injection haptics
- Anatomically accurate bone structure
- Visually identifiable landmarks
The Image Guided Cervical Spinal Injection Trainer enables trainees to develop a three-dimensional understanding of the procedures for successful spine interventions. They will learn to correlate imaging with the cervical spinal anatomy relevant to interventional pain procedures, to identify the target tissue and vulnerable structures using imaging and anatomic inspection, and to apply best practices to ensure patient safety during the interventional spine procedures.
BodyTextAccessories: See additional information for feature details.
Customers Who Bought This Also Bought