Vials, Polystyrene, with Snap Cap, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
PS Snap Cap Vials offer great rigidity and glass-like clarity.
- Lightweight
- Best for powders and non-liquid materials
Polyethylene caps 'snap' on and off quickly and easily. Great for storing bench top accessories and field sampling.
Customers Who Bought This Also Bought