ContactUsLinkComponent
Standard Bottles, Wide Mouth, Clear, Qorpak
Standard Bottles, Wide Mouth, Clear, Qorpak
# de catalogue 76332-922
Fournisseur:  Qorpak
Bottle Standard Wide Mouth Clr 16Oz CS24
Standard Bottles, Wide Mouth, Clear, Qorpak
# de catalogue 76332-922
Fournisseur:  Qorpak
Spécifications
  • Cap Size:
    63-400
  • Capacity:
    480 ml (16 oz.)
  • Description:
    Clear Standard Wide Mouths with Black Phenolic Pulp/Aluminum Foil Lined Cap
  • Cat. no.:
    76332-922
Spécifications
A propos de cet article
Clear, flint glass standard wide mouth bottle feature a wide opening making them the ideal general-purpose bottle.
  • Pulp/aluminum foil lined cap attached
  • Appropriate for non-acid, non-alkaline products, food applications, organic solvents, and mineral oils
Applications include pharmaceutical, chemical, environmental, and food packaging and storing.
Customers Who Bought This Also Bought