DURAN® Tamper Evident Red/Blue Screw Cap GL 45 with Linerless Plug Seal, Qorpak®
# de catalogue 76308-364
Fournisseur: Qorpak
Spécifications
- Cap Size:GL 45
- Color:Blue/Red
- Description:Tamper Evident Screw Cap
- Material:PP
- Cat. no.:76308-364
Spécifications
A propos de cet article
Seal GL 45 Media Bottles with secure cap.
- Tamper evident
- Autoclavable
- Linerless Plug Seal
Polypropylene material allows maximum temperature of 140 °C with minimum temperature of –40 °C.
Customers Who Bought This Also Bought