Polystyrene Spot Plates
# de catalogue 470313-468
Fournisseur: GSC International
Spécifications
- Description:Polystyrene Spot Plate
- Cat. no.:470313-468
Spécifications
A propos de cet article
Polystyrene spot plates have 12 depressions that are 20 mm in diameter by 10 mm in depth.
Overall plate dimension: 80×110 mm
Customers Who Bought This Also Bought