BoneClones® Stegosaurus Spike
# de catalogue 470027-634
Fournisseur: BONE CLONES, INC.
Small Business Enterprise
Spécifications
- Description:Stegosaurus Spike
- Cat. no.:470027-634
- Length:53.3 cm (21")
- Supplier No.:546226
Spécifications
A propos de cet article
Stegosaurus ungulatus, meaning 'roof lizard', dates from the late jurassic period and was first described by Marsh in 1879 based on bones found in wyoming.
- Casting from originals that present diverse osteological characteristics
- Each BoneClones® cast is hand-made from start to finish
- Rigorous standards are applied when selecting original specimens to ensure the best representation
- Proprietary, custom blend of polyurethane developed to capture and replicate minute detail
This 26' long, 6800 pound herbivore had a brain the size of a walnut. Triangular plates were positioned along its' back, where they may have served the purpose of temperature regulation or defense. The flexible tail featured eight spikes. This particular spike, found in Colorado in the mid 1990s, is one of the largest, most complete spikes around.
Customers Who Bought This Also Bought