Neurological Buck Hammer
Catalog # 470315-222
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Buck Hammer
- Cat. no.:470315-222
Specifications
About this item
Test reflexes and sensitivity.
- Stainless steel handle and head
- Integrated needle and brush
For testing reflex response and neurological sensitivity. This two sided hammer has a large and a small rounded head. In the handle is a brush for testing touch sensitivity. On the head of the hammer is a needle for testing for pain sensitivity. Note the needle included with this hammer is not intended to break the skin.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.