ADC® Adscope® 604 Pediatric Stethoscopes
# de catalogue 470315-678
Fournisseur: AMERICAN DIAGNOSTIC CORP CP
Spécifications
- Color:Black
- Description:Adscope 604
- Cat. no.:470315-678
Spécifications
A propos de cet article
Made specifically for pediatrics.
- Non chill bell and diaphragm
- Extra large bell for low frequency response
- Not made with natural rubber latex
The ADC Adscope 604 offers legendary acoustics, performance, and functionality for pediatric care. The combination pediatric chest piece offers both an ultra-sensitive diaphragm for greater amplification and crisper high frequency transmission as well as an extra deep bell for unsurpassed low-frequency response. The adjustable frequency (AFD) technology allows you to tune the diaphragm to further enhance high and low frequency response. The headset offers stainless-steel binaural fixed at a 15 degree angle for greater comfort and a reinforcing yoke molded into the 22" PVC tubing.
Customers Who Bought This Also Bought