VWR® A Balances with Calibration Certificate | Avantor
VWR® A Balances with Calibration Certificate
VWR® A Balances with Calibration Certificate
Supplier:  VWR International
Certificates
VWR® A Balances with Calibration Certificate
VWR® A Balances with Calibration Certificate
14 Options Available Below
Product Details & Documents
The VWR® A-Series balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • 2× RS 232, USB (A and B), and Wi-Fi communication interfaces
  • Five year warranty
  • Internal calibration
  • Easy-to-read, adjustable, backlit display
  • Tool-free assembly and diassembly of the 3-door draft shield on analytical models
The VWR® A-Series balances with calibration certificate include an ISO 17025:2017 certificate of calibration showing the full testing data using NIST-traceable weights. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-Series provide advanced features for any application. The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100,000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. The VWR® A-Series balances with calibration certificate are the answer for organizations demanding precision and performance with full traceability.
Certifications: Includes an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory issues calibration certificate. CE marked.
Ordering information: Optional accessories such as printer (10799-132), density determination kit and granite platform (10799-162) are available to order separately. The 0.01 and 0.1 mg readability models feature a draft shied (which can not be removed). The 0.001g readability models feature a removable, glass shield. All units are supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty. Accessories not included. Drop shipment only. Customer information required for calibration certificate.
Caution: Fragile and handle with care.
Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76458-520
  • 76458-518
  • 76458-522
  • 76458-524
  • 76458-526
  • 76458-528
  • 76329-734
  • 76329-736
  • 76329-738
  • 76329-740
  • 76329-742
  • 76329-960
  • 76458-530
  • 76329-962
  • Modèle:
  • VWR-205AC/CAL
  • VWR-105AC/CAL
  • VWR-160AC/CAL
  • VWR-220AC/CAL
  • VWR-310AC/CAL
  • VWR-520AC/CAL
  • VWR-363AC/CAL
  • VWR-603AC/CAL
  • VWR-1003AC/CAL
  • VWR-2102AC/CAL
  • VWR-4502AC/CAL
  • VWR-6002AC/CAL
  • VWR-8102AC/CAL
  • VWR-10102AC/CL
  • Operating temperature range:
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • 10...40 °C (50...104 °F)
  • Plug type:
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Nema 5-15R
  • Communication interface:
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • RS232 (2), USB-A, USB-B, and Wi-Fi
  • Voltage:
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
  • Input: 100 - 240 VAC, 50/60 Hz
Spécifications
Product Family Options
Analytical with Draft Shield
Informations sur le produit
  • Désignation
    Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • Semi-micro/analytical balance
    VWR-205AC/CAL
    82/220 g
    0.05 / 0.2 mg
    Ø 90 mm
    0.01 / 0.1 mg
    2 s
    0.02 mg (at 5% load)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,165.51
    $8,165.51
    Each
       $8,165.51
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-520
    Modèle
    VWR-205AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • Semi-micro balance
    VWR-105AC/CAL
    120 g
    0.07 mg
    Ø 90 mm
    0.01 mg
    2 s
    0.02 mg (at 5% load)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11,017.65
    $11,017.65
    Each
       $11,017.65
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-518
    Modèle
    VWR-105AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • Analytical balance
    VWR-160AC/CAL
    160 g
    0.2 mg
    Ø 100 mm
    0.1 mg
    2 s
    0.09 mg (at 5% load)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,046.11
    $4,046.11
    Each
       $4,046.11
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-522
    Modèle
    VWR-160AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • Analytical balance
    VWR-220AC/CAL
    220 g
    0.2 mg
    Ø 100 mm
    0.1 mg
    2 s
    0.09 mg (at 5% load)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,371.87
    $4,371.87
    Each
       $4,371.87
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-524
    Modèle
    VWR-220AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • Analytical balance
    VWR-310AC/CAL
    310 g
    0.2 mg
    Ø 100 mm
    0.1 mg
    2.5 s
    0.12 mg (at 5% load)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,697.64
    $4,697.64
    Each
       $4,697.64
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-526
    Modèle
    VWR-310AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • Analytical balance
    VWR-520AC/CAL
    520 g
    0.4 mg
    Ø 100 mm
    0.1 mg
    2.5 s
    0.12 mg (at 5% load)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,911.29
    $6,911.29
    Each
       $6,911.29
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-528
    Modèle
    VWR-520AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
Precision with Short, Removable Draft Shield
Informations sur le produit
  • Désignation
    Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • VWR-363AC/CAL
    360 g
    0.002 g
    128×128 mm
    0.001 g
    2 s
    0.0005 g (at 5% load)
    20.6W×33.3D×10H cm (8.11×13.1×3.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,455.03
    $2,455.03
    Prix promotionnel pour les nouveaux clients
    Each
       $2,455.03
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-734
    Modèle
    VWR-363AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • VWR-603AC/CAL
    600 g
    0.003 g
    128×128 mm
    0.001 g
    2 s
    0.0005 g (at 5% load)
    20.6W×33.3D×10H cm (8.11×13.1×3.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,046.08
    $4,046.08
    Each
       $4,046.08
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-736
    Modèle
    VWR-603AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • VWR-1003AC/CAL
    1000 g
    0.003 g
    128×128 mm
    0.001 g
    2 s
    0.0005 g (at 5% load)
    20.6W×33.3D×10H cm (8.11×13.1×3.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,697.65
    $4,697.65
    Each
       $4,697.65
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-738
    Modèle
    VWR-1003AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
Precision, No Draft Shield
Informations sur le produit
  • Désignation
    Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • VWR-2102AC/CAL
    2100 g
    0.02 g
    195×195 mm
    0.01 g
    1.5 s
    0.005 g (at 5% load)
    20.6W×33.3D×10H cm (8.11×13.1×3.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,044.34
    $3,044.34
    Each
       $3,044.34
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-740
    Modèle
    VWR-2102AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • VWR-4502AC/CAL
    4500 g
    0.02 g
    195×195 mm
    0.01 g
    1.5 s
    0.005 g (at 5% load)
    20.6W×33.3D×10H cm (8.11×13.1×3.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,068.76
    $3,068.76
    Each
       $3,068.76
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-742
    Modèle
    VWR-4502AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • VWR-6002AC/CAL
    6000 g
    0.03 g
    195×195 mm
    0.01 g
    1.5 s
    0.005 g (at 5% load)
    20.6W×33.3D×10H cm (8.11×13.1×3.9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,720.30
    $3,720.30
    Each
       $3,720.30
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-960
    Modèle
    VWR-6002AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • VWR-8102AC/CAL
    8100 g
    0.03 g
    195×195 mm
    0.01 g
    1.5 s
    0.005 g (at 5% load)
    20.6W×33.3D×10.7H cm (8.11×13.1×4.2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,772.56
    $4,772.56
    Each
       $4,772.56
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76458-530
    Modèle
    VWR-8102AC/CAL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz
  • VWR-10102AC/CL
    10100 g
    0.03 g
    195×195 mm
    0.01 g
    1.5 s
    0.005 g (at 5% load)
    20.6W×33.3D×10.7H cm (8.11×13.1×4.2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,512.07
    $5,512.07
    Each
       $5,512.07
    La quantité minimale de commande est de 1
    Spécifications:
    More
    Specifications
    Cat. No.
    76329-962
    Modèle
    VWR-10102AC/CL
    Operating temperature range
    10...40 °C (50...104 °F)
    Plug type
    Nema 5-15R
    Communication interface
    RS232 (2), USB-A, USB-B, and Wi-Fi
    Voltage
    Input: 100 - 240 VAC, 50/60 Hz