Loopster Digital Rotating Shaker, IKA
# de catalogue 76329-832
Fournisseur: IKA Works
Irregular Voltage
Spécifications
- Shaking motion:Rotating
- Description:Loopster Digital Rotating Shaker
- Model:Loopster Digital
- Max. load:2 kg
- Display:7 Segment LED
- Timer:1 Second to 5999 Minutes
- Cat. no.:76329-832
- W×D×H:300×300×360 mm
- Electrical:100/240 V, 50/60 Hz
- Speed Range:0 - 80 rpm
- Tilt Angle:0 - 90°
- Weight:3 kg
Spécifications
A propos de cet article
Digital rotator for smooth but effective mixing with adjustable speed. Ideal for biological samples, particularly blood samples.
- Usable at 4 to 50 °C in incubators
- Freely adjustable rotary angle
- Suitable for continuous operation
- Error code display
- Easy operation by foil keypad
- Timer: Countdown, adjustable from 1 second to 5999 minutes
Suitable for separating and mixing processes of powdered and liquid samples in Eppendorf tubes and Greiner tubes up to 50 ml. The easy removable attachments allow simple and quick cleaning of the unit of spilled samples.
Certifications: IP 21 protection class according to DIN EN 60529.
Customers Who Bought This Also Bought