Trayster Digital Overhead Shaker, IKA
# de catalogue 76329-838
Fournisseur: IKA Works
Spécifications
- Shaking motion:Overhead, Rotating
- W×D×H:460×140×180 mm
- Model:Trayster Digital
- Power input:16 W
- Speed:0 - 80 rpm
- Weight:3.28 kg
- Maximum Load:2 kg
- Voltage:100 – 240 V, 50/60 Hz
Spécifications
A propos de cet article
Digital overhead shaker with vertical rotation for smooth but effective mixing with infinitely adjustable speed. Ideal for biological samples, particularly blood samples.
- Usable at 4 to 50 °C in incubators
- Suitable for continuous operation
- Seven segment LED timer display with error code display
- Easy operation by foil keypad
- Counter display of shaking time
- Timer: Countdown, adjustable from one second to 5999 minutes
Suitable for separating and mixing processes of powdered and liquid samples in Eppendorf tubes and Greiner tubes up to 50 ml. The easy removable attachments allow simple and quick cleaning of the unit of spilled samples.
Certifications: IP 21 Protection class according to DIN EN 60529.
Customers Who Bought This Also Bought