VWR® Storage Bins with Lids for Mixed Use Storage
# de catalogue 76053-298
Fournisseur: VWR International
Spécifications
- Dimensions:Exterior: 2113/16L×11H×11"W
Interior: 211/2L×11H×103/4"W - Description:Bin Organizer with Lid
- Cat. no.:76053-298
Spécifications
A propos de cet article
Mixed use storage bins.
- Excellent for organizing –20* freezers and cold rooms
- Can accept a wide variety of tubes, bottles, boxes and so on
Constructed of corrosion resistant stainless steel.
Customers Who Bought This Also Bought