Rotational Viscometers, ROTAVISC, IKA
Fournisseur: IKA Works
Certificats
A propos de cet article
The ROTAVISC series determines the viscosity of liquids in all areas of application ranging from the laboratory to quality control. The four devices measure in different viscosity ranges.
- Easy to operate
- 4.3″ TFT display allows for an intuitive menu guidance
- Digital spirit level supports the correct set-up of the start-up condition
- Simple ramp function simplifies repetitive tasks
- Stepless speed adjustment
Regardless of a simple or demanding viscosity measurement - the ROTAVISC delivers rapid and accurate results.
The level of measurement accuracy of the ROTAVISC rotational viscometer for both Newtonian and non-Newtonian fluids is +/- 1 % of the measuring range. The reproducibility is +/- 0.2 %.
Certifications: IP 40 protection class according to DIN EN 60529.
Delivery: The scope of delivery includes a standard spindle set (SP1-SP4 or SP7-SP12), a protective bracket, temperature sensor and ROTASTAND stand.
Spécifications
- Catalog No:
- 76499-020
- 76499-022
- 76329-888
- Model:
- Hi-vi I
- Hi-vi II
- Lo-vi
- Accuracy:
- 1 %
- 1 %
- 1 %
- Speed range:
- 0.01 - 200 rpm
- 0.01 - 200 rpm
- 0.01 - 200 rpm
- Display:
- TFT, (4.3")
- TFT, (4.3")
- TFT, (4.3")
- Interfaces:
- USB and RS232 Interface
- USB and RS232 Interface
- USB and RS232 Interface
- Electrical:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- W×D×H:
- 351×372×629 mm
- 351×372×629 mm
- 351×372×629 mm