Hydrion® Lo Ion pH Test Kit, Micro Essential Laboratory
Catalogue # 60720-009
Fournisseur: Micro Essential
Numéro CAS:
Spécifications
- Description:Hydrion® Lo Ion Test Kit
- Cat. no.:60720-009
- Supplier No.:LI-5000
Spécifications
À propos de cet article
The Hydrion wide range Lo ion pH test kit is intended to test weakly buffered solutions and will give accurate pH readings at each full pH unit from pH 1.0-11.0. Estimates can be made to 0.5 ph units.
The color chart has pH matches at [1.0][2.0][3.0][4.0][5.0][6.0][7.0][8.0][9.0][10.0][11.0].
Delivery: Each kit contains a 15-foot roll of test paper, matching color chart and one(1) capped 4 ml. sample tube, packed in a polypropylene protective case.