ContactUsLinkComponent
Kiddiecart, 4-Drawer Cube, 26"
Kiddiecart, 4-Drawer Cube, 26"
# de catalogue 470317-022
Fournisseur:  KIDDER MFG. CO. LTD
KIDDIECART DRAWER CUBE
Kiddiecart, 4-Drawer Cube, 26"
# de catalogue 470317-022
Fournisseur:  KIDDER MFG. CO. LTD
Spécifications
  • Description:
    26" Kiddiecart, 4 Drawer Cube
  • Cat. no.:
    470317-022
Spécifications
A propos de cet article
This cart has been tried and tested in primary classes across the country and the response has been incredible.
  • 26×26" bamboo top (also 26" from floor)
  • 23×23" drawer housing
The perfect Mobile Maker/STEAM Cart has finally arrived for this specific age group. No more step stools or stands required for your primaries to reach the table top as this cart is the perfect height from the ground (26"). Accessibility is key for this age group to ensure optimal safety in your Maker environment. Four wheels and castors with lock and swivel make this cart ideal mobility around your classroom or school. There are four spacious drawers The robust, two foot square bamboo top is great for using your C-clamp, bench hook, mini hacksaw and more!
Customers Who Bought This Also Bought