VWR® T Balances, Legal-for-Trade (NTEP)
Fournisseur: VWR International
Certificats
A propos de cet article
The VWR® T-Series balances with NTEP certification feature state-of-the-art technology to provide the highest level of accuracy.
- Class I and II options
- Advanced features for weighing and documentation
- Internal calibration
- 2 x RS-232, wireless, ethernet, and USB (A & B) communication interfaces
- Touchless operation via IR proximity sensors
- 5-year warranty
These Class I (d=0.01 mg and 0.1 mg) and Class II (d=0.001 g and 0.01 g) balances offer the highest precision in legal-for-trade applications. The VWR® T-Series balances offer a variety of standard features, such as internal calibration, GLP procedures, adjustable ambient condition settings, 11 application modes, and multiple weighing units. The VWR® T-Series balances featuring an NTEP Certificate of Conformance to meet an organizations legal-for-trade requirements.
Certifications: CE Marked
BodyTextAccessories: Optional accessories such as printer (10799-132) and granite platform (10799-162) are available to order separately.
Ordering information: The 0.01 mg and 0.1 mg readability models feature a 3-door draft shield (which can not be removed). The 0.001 g readability models feature a removable glass draft shield. All units are supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Accessories not included.
Caution: Fragile. Handle with care.
Spécifications
- Catalog No:
- 76458-452
- 76458-454
- 76329-400
- 76458-442
- 76329-398
- 76458-448
- 76458-450
- 76458-444
- 76329-396
- 76458-446
- 76329-394
- 76329-392
- 76458-456
- Operating temperature range:
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- Plug type:
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Communication interface:
- Wireless, 2×RS232, USB (A and B), Ethernet
- Wireless, 2×RS232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS232, USB (A and B), Ethernet
- Wireless, 2×RS232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Wireless, 2×RS-232, USB (A and B), Ethernet
- Voltage:
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V
- 100 - 240V