Somso® Mammary Gland in Resting Position
# de catalogue 470102-548
Fournisseur: MARCUS SOMMER
Spécifications
- Description:Mammary Gland in Resting Position
- Height:25.5 cm
- Cat. no.:470102-548
- Length:18 cm
- Width:18 cm
- Supplier No.:MS 7
- No. of Parts:2
Spécifications
A propos de cet article
Divides into two parts
Five year warranty
- Somewhat enlarged, in SOMSO-Plast®. After drawings by Prof. Dr. Petry. Separates into two halves. On a stand with green base.
Ordering information: This item is a special order item. Available stock may be limited and additional lead time may be required.
Customers Who Bought This Also Bought