Econo™ Animal Restraints, Maryland Plastics
Supplier: Maryland Plastics
Certificates
About this item
These cages helps in holding animals from 30 to 500 g in size.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.