About this item
The A-series balances with NTEP certification feature state-of-the-art technology to provide the highest level of accuracy.
- Class I and II options
- Advanced features for weighing and documentation
- Internal calibration
- 2× RS232, USB (A and B), and Wi-Fi communication interfaces
- Easy-to-read, adjustable, backlit display
The VWR® A-Series Balances with NTEP Certification offer the highest level or precision in legal-for-trade applications. These Class I and Class II certified balances feature an NTEP Certificate of Conformance indicating the devices meet the requirements of Handbook 44. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR® A-Series balances provide advanced features for any application.
The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100000 weighings. The VWR® A-Series Balances with NTEP Certification are the answer for organizations demanding precision and performance in legal-for-trade applications.
AVBP-01 (10799-162): Granite platform
Specifications
- Catalog No:
- 76329-408
- 76329-406
- 76329-410
- Operating temperature range:
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- 10…40 °C (50…104 °F)
- Plug type:
- Nema 5-15R
- Nema 5-15R
- Nema 5-15R
- Communication interface:
- RS232 (2), USB (A and B)
- RS232 (2), USB-A, USB-B, and Wi-Fi
- RS232 (2), USB (A and B)
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V