Accessories for Laboratory Benchtop Autoclaves, Kinematica
Catalog # 97036-104
Supplier: Kinematica
Small Business Enterprise
Specifications
- Description:Adapter PT-DA / PTA (F / A) allows use of Old-Style Pt 10-35 Generators (Pta Style) with the New model Pt 10-35 Gt (Requires Chrome Lock Nut - 11090014)
- Cat. no.:97036-104
- Supplier No.:11035001