Rectangular and Square Cover Slips, Bellco Glass
Fournisseur: Bellco Glass
Certificats
A propos de cet article
These cover slips provides good quality, reliability, and better performance for all laboratory microscopy needs
- #1 thickness (0.13 to 0.16 mm)
- Packed 100 per case
Clear white borosilicate glass.
Customers Who Bought This Also Bought