About this item
This magnetic induction stirrer is motor-less, using advanced coil technology that produces the strongest magnetic coupling and has no moving parts making this unit 100% wear-free and maintenance free. The magnetic coil technology works on the inductive principle with alternative current (AC) as its driving force.
- No heat is generated after hours of continuous use
- Ideal for temperature sensitive samples and environments
- Quiet operation and low vibration
- The magnetic field driving the stir bar maintains continuous constant speeds for reproducible results
- Speed remains constant,even when load changes
- Rapid acceleration and braking, less than ten seconds
- An easy to read digital display precisely controls time (30 seconds to 60 minutes) and speed (50 to 2000 rpm)
- Durable ABS housing is corrosion resistant and easy to clean
This uses advanced coil technology resulting in the unit creating the strongest magnetic coupling, perfect for viscous solutions.
Once a program has started, the speed, cycle and time can be monitored. The stirring speed can be adjusted while a program is running.
Stirrer bars are coated with PTFE for chemical resistance. Stirrer modes: Single clockwise, single counter clockwise, auto reverse rotation.
Mixing Surface is 165 mm diameter, silicone mat is 160 mm in diameter.
Specifications
- Catalog No:
- 470316-810
- 470316-812
- Max. stirring capacity H₂O:
- 3 L plus
- 3 L plus
- Timer:
- 30 s - 60 min, variable, 1 second increments or continuous
- 30 s - 60 min, variable, 1 second increments or continuous