ChemRxnHub™ Systems, Jacketed, 1000 ml, Jacketed Lab Reactor, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.
Customers Who Bought This Also Bought