VWR® Cleanroom Wipes, 100% Polyester
# de catalogue 10835-982
Fournisseur: VWR International
Spécifications
- W×L:30.5×30.5 cm (12×12")
- Description:Cleanroom wipes, laser cut edges
- Material:Polyester, 100%
- Basis weight:115 g/m²
Spécifications
A propos de cet article
These are 100% continuous-filament, double-knit polyester fabric wipes.
- Free from silicone, amides, and DOP contamination
- Extremely low particle and fiber levels
- Laser cut edges
- Absorbant
Resistant to abrasion, providing maximum cleaning performance. Designed for cleaning surfaces and controlling spills in ISO Class 4 cleanroom environments.
Customers Who Bought This Also Bought