GTS Extreme Shippers, Cold Chain Technologies
Fournisseur: Cold Chain Technologies
Certificats
A propos de cet article
Whether shipping internationally or across the country the GTS Extreme shippers are built for extended duration transit and maintain strict temperature control for up to five days.
- Powered by award-winning Koolit® Advanced PCM GEL
- Error-free modular pack-out with single-step refrigerant conditioning
- Year-round performance in extreme temperatures
- Simplified ordering and inventory management
- Cost-effective for single-use shipments
- Advanced shipper design for increased payload, reduced weight, and fewer refrigerants needed
Recommended for use for high value commercial products, clinical trial products, and research and development shipments.
Customers Who Bought This Also Bought