UVP Pen-Ray® Field Packs, Analytik Jena
Supplier: Analytik Jena US
Certificates
About this item
Pen-Ray® Field Packs combine a Pen-Ray® UV light source and a DC power supply (with 9 V battery) in a compact, portable package.
- Field Packs are lightweight (0.3 lbs.) and small enough to fit in a pocket or be carried by the belt clip attached to the power supply
- Field Packs are available with a choice of options for customization or in three standard configurations
- Field Pack FP-L/S provides two interchangeable light sources (365 and 254 nm); a lamp connector is located on the outside of the power supply for this configuration only
- Other configurations come with the light source permanently connected to the power supply
- Quartz diameter: 6.5 mm
- Handle diameter: 9.5 mm
- Dimensions: Lighted: 44.5 mm /Overall: 108 mm
UVP Pen-Ray® Field Packs are lightweight and small, making them perfect for field use. Theys are available in three standard configurations, or may be customized with many different options. Two of the standard configurations come with the light source permanently connected to the power supply. UVP Field Pack model FP-L/S provides two interchangeable light sources, 365 and 254 nm, that are connected to the lamp on the outside of the power supply.
Customers Who Bought This Also Bought