Notrax® 974 Ergo Mat™ Grande™ Floor Mattings, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
Certificates
About this item
Ergo Mat™ Grande™ anti-fatigue floor mats provide comfort and ergonomic support with a full one inch thickness.
- Heavy-duty corrugated ribbed top surface
- Dense closed cell PVC foam base provides added cushioning comfort
- Corrugated ribbed texture provides traction and is easy to sweep clean
- Ribbed texture runs the length of the mat
- UniFusion™ bond guaranteed for the life of the mat
- Redstop™ non-slip backing to reduce mat slippage
- Beveled edges on all 4 sides
- Certified by the NFSI (National Floor Safety Institute)
The durable vinyl corrugated ribbed top surface provides directional traction to reduce slips and offers superior anti-fatigue support for applications where workers are required to stand for long periods of time. The top surface is combined with a dense closed cell foam base utilizing UniFusion™ technology, which virtually eliminates the possibility of de-lamination. All Notrax® laminate floor mats come standard with RedStop™, a uniquely engineered non-slip backing technology that eliminates slipping and sliding on smooth surfaces, such as tile, wood, marble, and treated concrete.
Material Laminate/Vinyl
Frequently Bought Together