Notrax® 539 WebTrax® Floor Mattings, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
Certificates
About this item
WebTrax® is a multi-functional vinyl floor mat that can be used in a variety of wet applications.
- Industrial grade extruded PVC
- Open construction of extruded vinyl has a wave design that creates a slip-resistant walking surface
- Oil-, grease-, and UV-resistant
- Performs well in different climates and adapts to any surface
- Free of: DOP, DMF, silicone, heavy metal, ozone depleting substances
- Factory compressed borders
The wave pattern design creates a slip-resistant walking surface that increases traction on wet and slippery floors. Antibacterial treated PVC material prevents the growth of fungi or bacteria while the open structure design allows for easy draining of liquids. The industrial grade PVC compound also provides greater resistance to caustic chemicals, oils, and greases.
Material PVC
Customers Who Bought This Also Bought