VWR® Horizontal MAXI MR Gel Electrophoresis System
# de catalogue 76314-682
Fournisseur: VWR International
Spécifications
- Gel size (W×L):20×25 cm
- Description:VWR® MAXI MR Horizontal Gel System
- Cat. no.:76314-682
- Buffer Volume:2000 ml
Spécifications
A propos de cet article
VWR® MAXI MR model includes a buffer circulation system, powered by the electrodes themselves.
The hydrogen bubbles produced at the cathode pass up a diagonally positioned tube connected to the two sides of the tank producing a gentle circulation of the buffer. The result is a no pH and ionic gradient - without the need of mechanical pumps.
Delivery: Includes integral buffer circulation, safety lid with retractable sheathed leads, UVT gel tray with gasketed end gates, three combs- one each 16 well, 24 well and 36 well (1.5 mm thick).
Customers Who Bought This Also Bought