Notrax® 109 Brush Step® Floor Mattings, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
Certificates
About this item
Brush Step® Matting features a lighter-weight double rib high/low design that performs the scraping function of the mat. The ribbed surface brushes, channels, and conceals dirt and debris. It is the perfect choice for light to medium traffic areas.
- Polypropylene carpet with vinyl backing
- 18 ounces of needle-punched yarn per square yard
- Low profile to perform in narrow clearance doorways
- Vinyl backing ensures minimal buckling and mat movement
- Ribs run the length of the mat
Ideal for indoor use in entranceways, lobbies, and all light to medium traffic areas.
Customers Who Bought This Also Bought