SP Wilmad-LabGlass ProCulture® Delong Style Shaker Flasks, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Ideal for growing and mixing of cell cultures, these Delong style shaker flasks have long necks to reduce splashing that could reach the closure.
- Type 1, Class A borosilicate glass construction
- Designed for polypropylene and stainless steel closures
- Polypropylene caps have ribbed interior for improved sealing
- Autoclavable
Baffles, spaced at 120 °C, allow for increased oxygen transfer to the culture medium when used in conjunction with a shaker.
Environmentally sustainable when compared to plastic.
Caution: Do not place on direct heat source.
Customers Who Bought This Also Bought