Media Bottles, Square, PET, TriForest
Fournisseur: TRIFOREST ENTERPRISES INC
Certificats
A propos de cet article
Bottles are ideal for the storage of culture media, reagents, serum, and biological buffers.
- Excellent gas barrier properties; reduced permeability to CO₂/O₂
- Sterility of 10⁻⁶ SAL
- Molded graduations for approximate volume reference
- Non-pyrogenic, nuclease-free, and non-cytotoxic
- Bottle and cap material are Certified Animal Derived Component-Free
Square polyethylene terephthalate bottles are available with or without high density polyethylene caps in shrink wrapped trays. Lightweight and shatter resistant, bottles are recommended for storage of all compatible solutions, biologicals, liquid culture media, and aqueous buffers.
Bottles are available with standard cap (PTCXXXXS-C), tamper evident cap (PTCXXXXS-A), or without cap (PTCXXXXS-N) for use with automated fill lines.
Certifications: Compliance with USP Class VI guidelines.
Customers Who Bought This Also Bought