Bulk Dispenser with Easy Access Front Tray, S-Curve
# de catalogue 10827-908
Fournisseur: S-Curve Technologies
Spécifications
- Dispenser type:Wall-mountable
- PPE type:Multi-use
- Description:EPD-1100 bulk dispenser with easy access front tray and lid, PETG
- Material:PETG
- Cat. no.:10827-908
- W×D×H:12×10×16"
- Supplier No.:EPD-1100LP
Spécifications
A propos de cet article
Dispenser for ear plugs, lens cleaners, bulk safety glasses, and more.
- The EPD-1100 is great for small bulk items
- Load the product in the top and the front tray offers easy access
- Our wall mounting bracket allows for easy removal for filling and cleaning
- Flame polished edges ensure a smooth finish
Customers Who Bought This Also Bought