Bulk Dispensers, Bouffant/Beard Cover/Hair Nets, No Front Opening, S-Curve
# de catalogue 10693-184
Fournisseur: S-Curve Technologies
Spécifications
- Dispenser type:Wall-mountable
- No. of compartments:1
- PPE type:Multi-use
- Description:BD-860 bulk dispenser with lid, PETG
- Material:PETG
- Cat. no.:10693-184
- W×D×H:10×6×10"
- Supplier No.:BD-860PETG
Spécifications
A propos de cet article
A smaller dispenser designed to hold bulk quantities of ear plugs, safety glasses, face masks, or beard covers.
- Available in acrylic or PETG
- Flame treated edges for a smooth finish
- Keyholes on back for wall mounting
Customers Who Bought This Also Bought